accessibility menu, dialog, popup

UserName

CAS RN 125995-13-3

CH3H3CCH3OO(R)(R)H3NOH3CCH3O

CAS RN 125995-13-3

IUPAC Name: 2-[(4R,6R)-6-[2-(tert-butoxy)-2-oxoethyl]-2,2-dimethyl-1,3-dioxan-4-yl]ethan-1-aminium
Synonyms: 2-[(4R,6R)-6-[2-(tert-butoxy)-2-oxoethyl]-2,2-dimethyl-1,3-dioxan-4-yl]ethanaminium
Molecular Weight (g/mol): 274.38
Molecular Formula: C14H28NO4
InChi Key: HWSHVKNLMBMKSR-GHMZBOCLSA-O
SMILES: CC(C)(C)OC(=O)C[C@H]1C[C@@H](CC[NH3+])OC(C)(C)O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results