accessibility menu, dialog, popup

UserName

CAS RN 1257651-17-4

BOOH3CCH3H3CH3CHNONHOOCH3CH3H3C

CAS RN 1257651-17-4

IUPAC Name: tert-butyl N-({[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamoyl}methyl)carbamate
Synonyms: tert-butyl N-({[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamoyl}methyl)carbamate
Molecular Weight (g/mol): 376.26
Molecular Formula: C19H29BN2O5
InChi Key: IYTPRTWBNLJZLK-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC(=O)NC1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results