CAS RN 1256580-46-7
CAS RN 1256580-46-7
IUPAC Name:
9-ethyl-6,6-dimethyl-8-[4-(morpholin-4-yl)piperidin-1-yl]-11-oxo-5H,6H,11H-benzo[b]carbazole-3-carbonitrile
Synonyms:
alectinib
Molecular Weight (g/mol):
482.63
Molecular Formula:
C30H34N4O2
InChi Key:
KDGFLJKFZUIJMX-UHFFFAOYSA-N
SMILES:
CCC1=C(C=C2C(=C1)C(=O)C1=C(NC3=CC(=CC=C13)C#N)C2(C)C)N1CCC(CC1)N1CCOCC1
1
–
2
of
2
results
2
Alectinib, MedChemExpress
MedChemExpress Alectinib (CH5424802) is a potent, selective, and orally available ALK inhibitor with an IC50 of 1.9 nM and a Kd value of 2.4 nM (in an ATP-competitive manner), and also inhibits ALK F1174L and ALK R1275Q with IC50s of 1 nM and 3.5 nM, respectively. Alectinib demonstrates effective central nervous system (CNS) penetration.