CAS RN 1255311-61-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Fluoro-4-(isopropylamino)benzeneboronic acid pinacol ester, 96%, Thermo Scientific™
CAS: 1255311-61-5 Synonym: 3-Fluoro-4-(isopropylamino)phenylboronic acid pinacol ester