CAS RN 125494-59-9
CAS RN 125494-59-9
IUPAC Name:
hydrogen {1-[1-(4-chlorophenyl)cyclobutyl]-3-methylbutyl}dimethylamine hydrate chloride
Synonyms:
hydrogen sibutramine hydrate chloride
Molecular Weight (g/mol):
334.33
Molecular Formula:
C17H29Cl2NO
InChi Key:
KFNNPQDSPLWLCX-UHFFFAOYNA-N
SMILES:
[H+].O.[Cl-].CC(C)CC(N(C)C)C1(CCC1)C1=CC=C(Cl)C=C1
1
–
3
of
3
results
1
Sibutramine Hydrochloride Monohydrate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Sibutramine Hydrochloride Monohydrate, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
3
Sibutramine hydrochloride monohydrate
CAS: 125494-59-9 Molecular Formula: C17H29Cl2NO Molecular Weight (g/mol): 334.33 InChI Key: KFNNPQDSPLWLCX-UHFFFAOYNA-N PubChem CID: 64765