accessibility menu, dialog, popup

UserName

CAS RN 125404-04-8

NNH2N(E/Z)

CAS RN 125404-04-8

IUPAC Name: 4-(2-phenylethenyl)pyrimidin-2-amine
Synonyms: 4-(2-phenylethenyl)pyrimidin-2-amine
Molecular Weight (g/mol): 197.24
Molecular Formula: C12H11N3
InChi Key: LNUXNUNUGIHCPA-UHFFFAOYSA-N
SMILES: NC1=NC=CC(C=CC2=CC=CC=C2)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results