CAS RN 1254036-71-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Nemiralisib, MedChemExpress
MedChemExpress Nemiralisib (GSK2269557 free base) is a potent and highly selective PI3Kδ inhibitor with a pKi of 9.9.