accessibility menu, dialog, popup

UserName

NNCOCl

CAS RN 125117-96-6

IUPAC Name: 2-chloro-5-isocyanatopyridine
Synonyms: 2-chloro-5-isocyanatopyridine
Molecular Weight (g/mol): 154.55
Molecular Formula: C6H3ClN2O
InChi Key: WHENXACISRCOHK-UHFFFAOYSA-N
SMILES: ClC1=NC=C(C=C1)N=C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results