CAS RN 124750-67-0
CAS RN 124750-67-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-[4-[(2-Butyl-4-chloro-5-formylimidazol-1-yl)methyl]phenyl]benzonitrile, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.