CAS RN 1246814-53-8
CAS RN 1246814-53-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Nepafenac-d5, MedChemExpress
MedChemExpress Nepafenac D5 (AHR-9434 D5) is the deuterium labeled Nepafenac, which is a selective COX-2 inhibitor.