CAS RN 124635-84-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Flurbiprofen Glyceryl Ester, TRC
CAS: 124635-84-3 Molecular Formula: C18H19FO4 Molecular Weight (g/mol): 318.34 Synonym: 2-Fluoro-α-methyl-[1,1'-Biphenyl]-4-acetic Acid 2,3-Dihydroxypropyl Ester,2,3-Dihydroxypropyl 2-(2-fluoro-[1,1'-biphenyl]-4-yl)propanoate SMILES: CC(C1=CC=C(C2=CC=CC=C2)C(F)=C1)C(OCC(O)CO)=O