CAS RN 124584-08-3
CAS RN 124584-08-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
MedChemExpress Nesiritide
MedChemExpress Nesiritide (Brain Natriuretic Peptide-32 human) is an agonist of natriuretic peptide receptors (NPRs), with Kd values of 7.3 and 13 pM for NPR-A and NPR-C, respectively.
2
MedChemExpress Nesiritide
MedChemExpress Nesiritide (Brain Natriuretic Peptide-32 human) is an agonist of natriuretic peptide receptors (NPRs), with Kd values of 7.3 and 13 pM for NPR-A and NPR-C, respectively.
3
MedChemExpress Nesiritide
MedChemExpress Nesiritide (Brain Natriuretic Peptide-32 human) is an agonist of natriuretic peptide receptors (NPRs), with Kd values of 7.3 and 13 pM for NPR-A and NPR-C, respectively.