CAS RN 1244640-48-9
CAS RN 1244640-48-9
IUPAC Name:
N-{2-[1-(3-fluorophenyl)-4-oxo-1,3,8-triazaspiro[4.5]decan-8-yl]ethyl}naphthalene-2-carboxamide hydrochloride
Synonyms:
N-{2-[1-(3-fluorophenyl)-4-oxo-1,3,8-triazaspiro[4.5]decan-8-yl]ethyl}naphthalene-2-carboxamide hydrochloride
Molecular Weight (g/mol):
482.98
Molecular Formula:
C26H28ClFN4O2
InChi Key:
RYLAMDMOILNBKN-UHFFFAOYSA-N
SMILES:
Cl.FC1=CC=CC(=C1)N1CNC(=O)C11CCN(CCNC(=O)C2=CC=C3C=CC=CC3=C2)CC1
1
–
1
of
1
results
1
Tocris Bioscience™ VU 0364739 hydrochloride
CAS: 1244640-48-9 Molecular Formula: C26H28ClFN4O2