CAS RN 123119-89-1
CAS RN 123119-89-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
3-(9H-Carbazol-4-yloxy)-1,2-propanediol, TRC
CAS: 123119-89-1 Molecular Formula: C15 H15 N O3 Molecular Weight (g/mol): 257.28 Synonym: 1,2-Propanediol, 3-(9H-carbazol-4-yloxy)-,3-(9H-Carbazol-4-yloxy)-1,2-propanediol,4-(2,3-Dihydroxy-1-propoxy)carbazole IUPAC Name: 3-(9H-carbazol-4-yloxy)propane-1,2-diol SMILES: OCC(O)COc1cccc2[nH]c3ccccc3c12
2
3-(9H-Carbazol-4-yloxy)propane-1,2-diol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.