CAS RN 1229453-99-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Acrizanib, MedChemExpress
MedChemExpress Acrizanib (LHA510) is a VEGFR-2 inhibitor, with an IC50 of 17.4 nM for BaF3-VEGFR-2.