accessibility menu, dialog, popup

UserName

CAS RN 122520-85-8

H2NOOHHON(E)

CAS RN 122520-85-8

IUPAC Name: (2E)-2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enamide
Synonyms: (2E)-2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enamide
Molecular Weight (g/mol): 204.19
Molecular Formula: C10H8N2O3
InChi Key: USOXQZNJFMKTKJ-XVNBXDOJSA-N
SMILES: NC(=O)C(=C\C1=CC=C(O)C(O)=C1)\C#N
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)