CAS RN 12236-82-7
CAS RN 12236-82-7
IUPAC Name:
1-amino-4-{[4-({4-chloro-6-[(3-sulfophenyl)amino]-1,3,5-triazin-2-yl}amino)-3-sulfophenyl]amino}-9,10-dioxo-9,10-dihydroanthracene-2-sulfonic acid
Synonyms:
1-amino-4-{[4-({4-chloro-6-[(3-sulfophenyl)amino]-1,3,5-triazin-2-yl}amino)-3-sulfophenyl]amino}-9,10-dioxoanthracene-2-sulfonic acid
Molecular Weight (g/mol):
774.15
Molecular Formula:
C29H20ClN7O11S3
InChi Key:
JQYMGXZJTCOARG-UHFFFAOYSA-N
SMILES:
NC1=C(C=C(NC2=CC=C(NC3=NC(Cl)=NC(NC4=CC=CC(=C4)S(O)(=O)=O)=N3)C(=C2)S(O)(=O)=O)C2=C1C(=O)C1=CC=CC=C1C2=O)S(O)(=O)=O
1
–
2
of
2
results
1
MedChemExpress Procion Blue HB
MedChemExpress Procion Blue HB (Reactive Blue 2) is a purinergic antagonist.
2
MedChemExpress Procion Blue HB
MedChemExpress Procion Blue HB (Reactive Blue 2) is a purinergic antagonist.