CAS RN 1222689-74-8
CAS RN 1222689-74-8
IUPAC Name:
9-[4-amino-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-3H-purin-6-one
Synonyms:
9-[4-amino-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
Molecular Weight (g/mol):
251.25
Molecular Formula:
C10H13N5O3
InChi Key:
CHWLAIBBKPETPZ-UHFFFAOYNA-N
SMILES:
NC1CC(OC1CO)N1C=NC2=C1NC=NC2=O