CAS RN 1221186-53-3
CAS RN 1221186-53-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Promotions Available
Tocris Bioscience™ TEPP 46
Tocris Bioscience™ TEPP 46
Potent and selective pyruvate kinase M2 (PKM2) allosteric activator
2
TEPP-46, MedChemExpress
MedChemExpress TEPP-46 (ML-265) is a potent and selective pyruvate kinase M2 (PKM2) activator with an AC50 of 92 nM, showing little or no effect on PKM1, PKL and PKR.