accessibility menu, dialog, popup

UserName

CAS RN 120737-77-1

CH3NNOOH3CH3CCH3

CAS RN 120737-77-1

IUPAC Name: tert-butyl 4-benzyl-2-methylpiperazine-1-carboxylate
Synonyms: tert-butyl 4-benzyl-2-methylpiperazine-1-carboxylate
Molecular Weight (g/mol): 290.41
Molecular Formula: C17H26N2O2
InChi Key: ZPDZNQZBTLCTDV-UHFFFAOYNA-N
SMILES: CC1CN(CC2=CC=CC=C2)CCN1C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results