CAS RN 1207064-61-6
CAS RN 1207064-61-6
IUPAC Name:
{2-[3-(3-fluorobenzenesulfonyl)-1H-pyrrolo[2,3-b]pyridin-1-yl]ethyl}dimethylamine dihydrochloride
Synonyms:
{2-[3-(3-fluorobenzenesulfonyl)pyrrolo[2,3-b]pyridin-1-yl]ethyl}dimethylamine dihydrochloride
Molecular Weight (g/mol):
420.32
Molecular Formula:
C17H20Cl2FN3O2S
InChi Key:
BAOHMLBVCLITHA-UHFFFAOYSA-N
SMILES:
Cl.Cl.CN(C)CCN1C=C(C2=CC=CN=C12)S(=O)(=O)C1=CC=CC(F)=C1