missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ WAY 208466 dihydrochloride
GREENER_CHOICE

Product Code. 18074452
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18074452 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18074452 Supplier Tocris Bioscience™ Supplier No. 3904/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective 5-HT6 agonist

High affinity, selective 5-HT6 agonist (EC50 = 7.3 nM at the human 5-HT6 receptor). Elevates cortical GABA levels in vivo in rat frontal cortex. Exhibits antidepressant and anxiolytic-like effects.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1207064-61-6
Quantity 10 mg
Molecular Formula C17H20Cl2FN3O2S
Formula Weight 420.33
Purity >99%
Molecular Weight (g/mol) 420.324
Chemical Name or Material WAY 208466 dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.