CAS RN 1206731-57-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
WT-161, MedChemExpress
MedChemExpress WT-161 is a potent and selective HDAC6 inhibitor with an IC50 of 0.40 nM. WT-161 also inhibits metallo-β-lactamase domain-containing protein 2 (MBLAC2).