accessibility menu, dialog, popup

UserName

CAS RN 1206593-24-9

CH3OOHFFF

CAS RN 1206593-24-9

IUPAC Name: 4-ethoxy-3-(trifluoromethyl)phenol
Synonyms: 4-ethoxy-3-(trifluoromethyl)phenol
Molecular Weight (g/mol): 206.16
Molecular Formula: C9H9F3O2
InChi Key: KTAPXNQLDCIIAO-UHFFFAOYSA-N
SMILES: CCOC1=C(C=C(O)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results