CAS RN 120635-25-8
CAS RN 120635-25-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Mofegiline hydrochloride, MedChemExpress
MedChemExpress Mofegiline hydrochloride (MDL72974A) is a potent and selective enzyme-activated irreversible inhibitor of MAO-B; shows marked selectivity for the B form (IC50 = 680 and 3.6 nM for MAO-A and MAO-B).