accessibility menu, dialog, popup

UserName

CAS RN 1206163-56-5

BOOCH3CH3CH3CH3N

CAS RN 1206163-56-5

IUPAC Name: 1-benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
Synonyms: 1-benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indole
Molecular Weight (g/mol): 333.24
Molecular Formula: C21H24BNO2
InChi Key: ZJVGZQOSLAZPOI-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C2N(CC3=CC=CC=C3)C=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results