CAS RN 120406-37-3
CAS RN 120406-37-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Jionoside B1, MedChemExpress
MedChemExpress Jionoside B1 is a phenylpropanoid isolated from herbs of Eriophyton wallichii.