CAS RN 1197194-61-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Endoxifen E-isomer hydrochloride, MedChemExpress
MedChemExpress Endoxifen E-isomer hydrochloride (E-Endoxifen hydrochloride), an E-isomer of Endoxifen, is an impurity in Endoxifen Z-isomer drug substance. Endoxifen E-isomer hydrochloride exhibits antiestrogenic effects.