CAS RN 119637-67-1
CAS RN 119637-67-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Moguisteine, MedChemExpress
MedChemExpress Moguisteine(BBR-2173) is a novel peripheral non-narcotic antitussive drug.