CAS RN 1194961-19-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
PRT 060318, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
PRT 060318, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
PRT 060318, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
PRT-060318, MedChemExpress
MedChemExpress PRT-060318 (PRT318) is a novel selective inhibitor of the tyrosine kinase Syk with an IC50 of 4 nM.