CAS RN 118584-50-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
24,25-Dihydroxy Vitamin D2-d3 (Mixture of Diastereomers), TRC
CAS: 118584-50-2 Molecular Formula: C28 2H3 H41 O3 Molecular Weight (g/mol): 431.67 Synonym: (4E,6R)-2-Methyl-3-(methyl-d3)-6-[(1R,3aS,4E,7aR)-octahydro-4-[(2Z)-2-[(5S)-5-hydroxy-2-methylenecyclohexylidene]ethylidene]-7a-methyl-1H-inden-1-yl]-4-heptene-2,3-diol,(3β,5Z,7E,22E,24Ξ)-9,10-Secoergosta-5,7,10(19),22-tetraene-3,24,25-triol-d3 IUPAC Name: (E,6R)-6-[(1R,3aS,4E,7aR)-4-[(2Z)-2-[(5S)-5-hydroxy-2-methylidenecyclohexylidene]ethylidene]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-1-yl]-2-methyl-3-(trideuteriomethyl)hept-4-ene-2,3-diol SMILES: [2H]C([2H])([2H])C(O)(\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@@H](O)CCC3=C)\CCC[C@]12C)C(C)(C)O