CAS RN 118525-40-9
CAS RN 118525-40-9
IUPAC Name:
3,5,7-trihydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-en-1-yl)-4H-chromen-4-one
Synonyms:
3,5,7-trihydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-en-1-yl)chromen-4-one
Molecular Weight (g/mol):
368.39
Molecular Formula:
C21H20O6
InChi Key:
TUUXBSASAQJECY-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C(CC=C(C)C)=C2O1
1
–
1
of
1
results
1
Icaritin, MedChemExpress
MedChemExpress Icaritin (Anhydroicaritin) is a prenylflavonoid derivative from Epimedium Genusis and potently inhibits proliferation of K562 cells (IC50 of 8 μM) and primary CML cells (IC50 of 13.4 μM for CML-CP and 18 μM for CML-BC). Icaritin can regulate MAPK/ERK/JNK and JAK2/STAT3 /AKT signalings, also enhances osteogenesis[3.