accessibility menu, dialog, popup

UserName

H3COHNOOCH3H3CCH3OOH

CAS RN 117833-18-8

IUPAC Name: 4-(tert-butoxy)-2-acetamido-4-oxobutanoic acid
Synonyms: 4-(tert-butoxy)-2-acetamido-4-oxobutanoic acid
Molecular Weight (g/mol): 231.25
Molecular Formula: C10H17NO5
InChi Key: NKNPTCBHHPHSEA-UHFFFAOYNA-N
SMILES: CC(=O)NC(CC(=O)OC(C)(C)C)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results