CAS RN 1173203-26-3
CAS RN 1173203-26-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
4-[[3-(1-Methylethyl)-2-oxo-5-oxazolidinyl]methoxy]benzaldehyde, TRC
CAS: 1173203-26-3 Molecular Formula: C14 H17 N O4 Molecular Weight (g/mol): 263.29 Synonym: 4-[(3-Isopropyl-2-oxo-1,3-oxazolidin-5-yl)methoxy]benzaldehyde,Bisoprolol Fumarate Imp. T (EP),Bisoprolol Oxazolidinone-benzaldehyde IUPAC Name: 4-[(2-oxo-3-propan-2-yl-1,3-oxazolidin-5-yl)methoxy]benzaldehyde SMILES: CC(C)N1CC(COc2ccc(C=O)cc2)OC1=O
2
4-[(3-Isopropyl-2-oxo-1,3-oxazolidin-5-yl)methoxy]benzaldehyde (Bisoprolol Oxazolidinone-benzaldehyde), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.