accessibility menu, dialog, popup

UserName

CAS RN 1165936-03-7

BOOH3CCH3H3CH3CCH3F

CAS RN 1165936-03-7

IUPAC Name: 2-(2-fluoro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(2-fluoro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 236.09
Molecular Formula: C13H18BFO2
InChi Key: MKWDECBHRFNFGY-UHFFFAOYSA-N
SMILES: CC1=CC(F)=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results