CAS RN 1159977-09-9
CAS RN 1159977-09-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac Benzyl Phenylephrine(Phenylephrine Impurity D), TRC
CAS: 1159977-09-9 Molecular Formula: C16H19NO2 Molecular Weight (g/mol): 257.33 Synonym: Phenylephrine Impurity D,3-Hydroxy-α-[[methyl(phenylmethyl)amino]methyl]benzenemethanol; IUPAC Name: 3-[2-[benzyl(methyl)amino]-1-hydroxy-ethyl]phenol SMILES: CN(CC(O)c1cccc(O)c1)Cc2ccccc2