CAS RN 1140964-99-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
E7449, MedChemExpress
MedChemExpress E7449 is a potent PARP1 and PARP2 inhibitor and also inhibits TNKS1 and TNKS2, with IC50s of 2.0, 1.0, ∼50 and ∼50 nM for PARP1, PARP2, TNKS1 and TNKS2, respectively, using 32P-NAD+ as substrate.