accessibility menu, dialog, popup

UserName

CAS RN 1132787-68-8

CH3H3COHONHSOOCl

CAS RN 1132787-68-8

IUPAC Name: 2-(3-chlorobenzenesulfonamido)-4-methylpentanoic acid
Synonyms: 2-(3-chlorobenzenesulfonamido)-4-methylpentanoic acid
Molecular Weight (g/mol): 305.77
Molecular Formula: C12H16ClNO4S
InChi Key: KJQWUNLTCVDQQZ-UHFFFAOYNA-N
SMILES: CC(C)CC(NS(=O)(=O)C1=CC(Cl)=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)