accessibility menu, dialog, popup

UserName

H3COFFOOHF

CAS RN 112811-65-1

IUPAC Name: 2,4,5-trifluoro-3-methoxybenzoic acid
Synonyms: 2,4,5-trifluoro-3-methoxybenzoic acid
Molecular Weight (g/mol): 206.12
Molecular Formula: C8H5F3O3
InChi Key: YVJHZWWMKFQKDC-UHFFFAOYSA-N
SMILES: COC1=C(F)C(=CC(F)=C1F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results