accessibility menu, dialog, popup

UserName

ON

CAS RN 112598-77-3

IUPAC Name: 3-(furan-2-yl)benzonitrile
Synonyms: 3-(furan-2-yl)benzonitrile
Molecular Weight (g/mol): 169.18
Molecular Formula: C11H7NO
InChi Key: BUZQKXZXWYCNJK-UHFFFAOYSA-N
SMILES: N#CC1=CC(=CC=C1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results