accessibility menu, dialog, popup

UserName

H3CNCH3CH3

CAS RN 112-75-4

IUPAC Name: dimethyl(tetradecyl)amine
Synonyms: dimethyl myristamine
Molecular Weight (g/mol): 241.46
Molecular Formula: C16H35N
InChi Key: SFBHPFQSSDCYSL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCN(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results