accessibility menu, dialog, popup

UserName

CAS RN 111524-95-9

OOHNHOO

CAS RN 111524-95-9

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2-phenylacetic acid
Synonyms: {[(9H-fluoren-9-ylmethoxy)carbonyl]amino}(phenyl)acetic acid
Molecular Weight (g/mol): 373.41
Molecular Formula: C23H19NO4
InChi Key: PCJHOCNJLMFYCV-UHFFFAOYNA-N
SMILES: OC(=O)C(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)