CAS RN 109904-25-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
4,5,6,7-Tetrahydro-5-(triphenylmethyl)thieno[3,2-c]pyridine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4,5,6,7-Tetrahydro-5-(triphenylmethyl)thieno[3,2-c]pyridine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4,5,6,7-Tetrahydro-5-(triphenylmethyl)thieno[3,2-c]pyridine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.