accessibility menu, dialog, popup

UserName

NaH3C(S)(S)(R)(S)(R)H3COSOOO(S)O

CAS RN 1099-87-2

IUPAC Name: sodium (3aS,3bR,7S,9aR,9bS,11aS)-9a,11a-dimethyl-1-oxo-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl sulfate
Synonyms: sodium dehydroepiandrosterone sulfate
Molecular Weight (g/mol): 390.47
Molecular Formula: C19H27NaO5S
InChi Key: GFJWACFSUSFUOG-ZJTJBYBXSA-M
SMILES: [Na+].C[C@]12CC[C@H]3[C@@H](CC=C4C[C@H](CC[C@]34C)OS([O-])(=O)=O)[C@@H]1CCC2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results