accessibility menu, dialog, popup

UserName

CAS RN 1097079-85-0

CH3H3CNOCl

CAS RN 1097079-85-0

IUPAC Name: N-benzyl-4-(chloromethyl)-N-(propan-2-yl)benzamide
Synonyms: N-benzyl-4-(chloromethyl)-N-isopropylbenzamide
Molecular Weight (g/mol): 301.81
Molecular Formula: C18H20ClNO
InChi Key: BLONTFYZTPFISN-UHFFFAOYSA-N
SMILES: CC(C)N(CC1=CC=CC=C1)C(=O)C1=CC=C(CCl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results