CAS RN 1093403-33-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
SRT 2104, MedChemExpress
MedChemExpress SRT 2104 is a first-in-class, highly selective and brain-permeable activator of the NAD+ dependent deacetylase Sirt1, increases Sirt1 protein, but shows no effect on Sirt1 mRNA. Used in the research of diabetes mellitus and Huntington’s disease.