CAS RN 1093119-54-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MKC3946, MedChemExpress
MedChemExpress MKC3946 is a potent IRE1α inhibitor, used for cancer research.