CAS RN 1092461-27-2
IUPAC Name:
3-ethoxy-2,6-difluorobenzoyl chloride
Synonyms:
3-ethoxy-2,6-difluorobenzoyl chloride
Molecular Weight (g/mol):
220.6
Molecular Formula:
C9H7ClF2O2
InChi Key:
XGDCQLWWBIGNFG-UHFFFAOYSA-N
SMILES:
CCOC1=C(F)C(C(Cl)=O)=C(F)C=C1
1
–
1
of
1
results
1
3-Ethoxy-2,6-difluorobenzoyl chloride, JRD, 97%, Thermo Scientific™
CAS: 1092461-27-2 Molecular Formula: C9H7ClF2O2 Molecular Weight (g/mol): 220.60 MDL Number: MFCD11519369 InChI Key: XGDCQLWWBIGNFG-UHFFFAOYSA-N Synonym: 3-ethoxy-2,6-difluorobenzoyl chloride, jrd PubChem CID: 57361367 IUPAC Name: 3-ethoxy-2,6-difluorobenzoyl chloride SMILES: CCOC1=C(F)C(C(Cl)=O)=C(F)C=C1