CAS RN 109089-77-2
IUPAC Name:
3-(2-methoxy-5-methylphenyl)-3-phenylpropanoic acid
Synonyms:
3-(2-methoxy-5-methylphenyl)-3-phenylpropanoic acid
Molecular Weight (g/mol):
270.33
Molecular Formula:
C17H18O3
InChi Key:
KSLPLDLDQANKOD-UHFFFAOYNA-N
SMILES:
COC1=C(C=C(C)C=C1)C(CC(O)=O)C1=CC=CC=C1
1
–
3
of
3
results
1
3-(2-Methoxy-5-methylphenyl)-3-phenyl-propanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-(2-Methoxy-5-methylphenyl)-3-phenyl-propanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-(2-Methoxy-5-methylphenyl)-3-phenyl-propanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.