CAS RN 1087784-16-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
2-Chloro-1-{1H,2H,3H,4H,5H-pyrido[4,3-b]indol-2-yl}ethan-1-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Chloro-1-{1H,2H,3H,4H,5H-pyrido[4,3-b]indol-2-yl}ethan-1-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Chloro-1-{1H,2H,3H,4H,5H-pyrido[4,3-b]indol-2-yl}ethan-1-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.